![Deacylgymnemic acid phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/148/310/ad83c2ba-baff-4bc1-bd71-04a21170bc2c/640/ad83c2ba-baff-4bc1-bd71-04a21170bc2c.png)
Deacylgymnemic acid
![Deacylgymnemic acid phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/148/310/ad83c2ba-baff-4bc1-bd71-04a21170bc2c/640/ad83c2ba-baff-4bc1-bd71-04a21170bc2c.png)
Synonym(s):
(3β,4α,16β-21β,22α)-16,21,22,23,28-Pentahydroxyolean-12-en-3-yl β-D-glucopyranosiduronic acid, 3-O-β-D-Glucuronopyranosyl gymnemagenin
Empirical Formula (Hill Notation):
C36H58O12
CAS Number:
Molecular Weight:
682.84
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing