Skip to Content
Merck
D-(+)-Melezitose hydrate ≥97% (HPLC)

D-(+)-Melezitose hydrate

D-(+)-Melezitose hydrate ≥97% (HPLC)
Synonym(s):
α-D-Glc-[1→3]-β-D-Fru-[2→1]-α-D-Glc
Empirical Formula (Hill Notation):
C18H32O16 · xH2O
CAS Number:
Molecular Weight:
504.44 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing