![(+)-Biotin 4-nitrophenyl ester 98%](/deepweb/assets/sigmaaldrich/product/structures/358/909/f8e11efa-6f75-4770-9460-194daafc6ab7/640/f8e11efa-6f75-4770-9460-194daafc6ab7.png)
(+)-Biotin 4-nitrophenyl ester
![(+)-Biotin 4-nitrophenyl ester 98%](/deepweb/assets/sigmaaldrich/product/structures/358/909/f8e11efa-6f75-4770-9460-194daafc6ab7/640/f8e11efa-6f75-4770-9460-194daafc6ab7.png)
Synonym(s):
4-Nitrophenyl (+)-biotinate, d-Biotin p-nitrophenyl ester, BNP
Empirical Formula (Hill Notation):
C16H19N3O5S
CAS Number:
Molecular Weight:
365.40
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing