Skip to Content
Merck
Cytosine β-D-arabinofuranoside hydrochloride crystalline

Cytosine β-D-arabinofuranoside hydrochloride

Cytosine β-D-arabinofuranoside hydrochloride crystalline
Synonym(s):
1-(β-D-Arabino­furanosyl)­cytosine hydrochloride, Ara-C hydrochloride, Arabinocytidine hydrochloride, Arabinosylcytosine hydrochloride, Cytarabine hydrochloride, Cytosine arabinoside hydrochloride
Empirical Formula (Hill Notation):
C9H13N3O5 · HCl
CAS Number:
Molecular Weight:
279.68
EC Number:

Products

Brand
Biological Source
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing