Cyclohexylmethanol
Synonym(s):
Cyclohexylmethanol, Cyclohexanemethanol, (Hydroxymethyl)cyclohexane, Cyclohexyl methanol, Hexahydrobenzyl alcohol, NSC 5288
Linear Formula:
C6H11CH2OH
CAS Number:
Molecular Weight:
114.19
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3