Cumene hydroperoxide
Synonym(s):
α,α-Dimethylbenzyl hydroperoxide
Linear Formula:
C6H5C(CH3)2OOH
CAS Number:
Molecular Weight:
152.19
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing