![Crotamiton for peak identification CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/234/628/e90f0482-91e0-449d-b5bc-51d99d2879d2/640/e90f0482-91e0-449d-b5bc-51d99d2879d2.png)
Crotamiton for peak identification
![Crotamiton for peak identification CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/234/628/e90f0482-91e0-449d-b5bc-51d99d2879d2/640/e90f0482-91e0-449d-b5bc-51d99d2879d2.png)
Synonym(s):
N-Ethyl-o-crotonotoluidide, N-Ethyl-N-(2-methylphenyl)-2-butenamide, Crotamiton
Linear Formula:
CH3CH=CHCON(C2H5)C6H4CH3
CAS Number:
Molecular Weight:
203.28
Products
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Application
Markush Class
Markush Group
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing