Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Corning
®
384 Well Deep Well Plate
Products
Product Category
plates (4)
Color
Manufacturer
Binding Type
Volume (µL)
Wells
Color
Package Size
Sterility
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
CLS3342
Corning
®
384 Well Deep Well Plate,
clear polypropylene, V-bottom, sterile, deep well, lid: no, pack of 5
Expand
View Pricing
CLS3347
Corning
®
384 Well Deep Well Plate,
clear polypropylene, V-bottom, non-sterile, deep well, lid: no, pack of 10
Expand
View Pricing
CLS3964
Corning
®
384 Well Deep Well Plate,
clear polypropylene, round bottom, sterile, deep well, lid: no, pack of 5
Expand
View Pricing
CLS3965
Corning
®
384 Well Deep Well Plate,
clear polypropylene, round bottom, non-sterile, deep well, lid: no, pack of 5
Expand
View Pricing