Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cobalt/Silicon/Molybdenum/Iron/Boron/Niobium
Products
Manufacturer
Physical Form
Length (m)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
GF24941268
Cobalt/Silicon/Molybdenum/Iron/Boron/Niobium,
foil, Co 78%/Si 9%/Mo 5%/Fe 4%/B 2%/Nb 2%, thickness 0.023 mm, length 20 m, coil width 20 mm
Expand
View Pricing
GF08815787
Cobalt/Silicon/Molybdenum/Iron/Boron/Niobium,
foil, Co 78%/Si 9%/Mo 5%/Fe 4%/B 2%/Nb 2%, thickness 0.023 mm, length 10 m, coil width 20 mm
Expand
View Pricing
GF28575995
Cobalt/Silicon/Molybdenum/Iron/Boron/Niobium,
foil, Co 78%/Si 9%/Mo 5%/Fe 4%/B 2%/Nb 2%, thickness 0.023 mm, length 1 m, coil width 20 mm
Expand
View Pricing
GF34822504
Cobalt/Silicon/Molybdenum/Iron/Boron/Niobium,
foil, Co 78%/Si 9%/Mo 5%/Fe 4%/B 2%/Nb 2%, thickness 0.023 mm, length 2 m, coil width 20 mm
Expand
View Pricing
GF96979291
Cobalt/Silicon/Molybdenum/Iron/Boron/Niobium,
foil, Co78%/Si 9%/Mo 5%/Fe 4%/B 2%/Nb 2%, thickness 0.023 mm, length 5 m, coil width 20 mm
Expand
View Pricing