![Clotrimazole Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/348/828/dde1c39d-b257-4e70-9f2a-7b1f656f3a0c/640/dde1c39d-b257-4e70-9f2a-7b1f656f3a0c.jpg)
Clotrimazole related compound A
![Clotrimazole Related Compound A Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/348/828/dde1c39d-b257-4e70-9f2a-7b1f656f3a0c/640/dde1c39d-b257-4e70-9f2a-7b1f656f3a0c.jpg)
Synonym(s):
(o-Chlorophenyl)diphenylmethanol
Empirical Formula (Hill Notation):
C19H15ClO
CAS Number:
Molecular Weight:
294.77
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2