![Clonidine Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/418/900/0b60150b-12bd-4a75-9b7c-63dbd3e2ec9a/640/0b60150b-12bd-4a75-9b7c-63dbd3e2ec9a.jpg)
Clonidine Related Compound A
![Clonidine Related Compound A certified reference material, pharmaceutical secondary standard](/deepweb/assets/sigmaaldrich/product/images/418/900/0b60150b-12bd-4a75-9b7c-63dbd3e2ec9a/640/0b60150b-12bd-4a75-9b7c-63dbd3e2ec9a.jpg)
Synonym(s):
1-Acetyl-2-[(2,6-dichlorophenyl)amino]-4,5-dihydro-1H-imidazole
Empirical Formula (Hill Notation):
C11H11Cl2N3O
CAS Number:
Molecular Weight:
272.13
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2