Skip to Content
Merck
vibrant-m

Citalopram Related Compound H

vibrant-m
Synonym(s):
5-Bromo-1-(4-fluorophenyl)-1,3-dihydro-N,N-dimethyl-1-isobenzofuranpropanamine hydrobromide
Empirical Formula (Hill Notation):
C19H21BrFNO · HBr
CAS Number:
Molecular Weight:
459.19

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1