Skip to Content
Merck
cis-5,8,11,14,17-Eicosapentaenoic acid sodium salt ≥99% (capillary GC)

cis-5,8,11,14,17-Eicosapentaenoic acid sodium salt

cis-5,8,11,14,17-Eicosapentaenoic acid sodium salt ≥99% (capillary GC)
Linear Formula:
CH3(CH2CH=CH)5(CH2)3CO2Na
CAS Number:
Molecular Weight:
324.43

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing