Skip to Content
Merck
cis-3-Hexenyl lactate natural, ≥97%, FG

cis-3-Hexenyl lactate

cis-3-Hexenyl lactate natural, ≥97%, FG
Linear Formula:
CH3CH(OH)CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
172.22
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Purity
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing