Skip to Content
Merck
Ciprofibrate European Pharmacopoeia (EP) Reference Standard

Ciprofibrate

Ciprofibrate European Pharmacopoeia (EP) Reference Standard
Synonym(s):
2-[p-(2,2-Dichlorocyclopropyl)phenoxy]-2-methylpropanoic acid, Ciprofibrate
Empirical Formula (Hill Notation):
C13H14Cl2O3
CAS Number:
Molecular Weight:
289.15

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3