![Chlorpromazine impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/399/404/d45c4a04-9abe-496b-848d-0ea03da603c6/640/d45c4a04-9abe-496b-848d-0ea03da603c6.png)
Chlorpromazine impurity E
![Chlorpromazine impurity E European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/399/404/d45c4a04-9abe-496b-848d-0ea03da603c6/640/d45c4a04-9abe-496b-848d-0ea03da603c6.png)
Synonym(s):
2-Chlorophenothiazine, 2-Chlorothiodiphenylamine
Empirical Formula (Hill Notation):
C12H8ClNS
CAS Number:
Molecular Weight:
233.72
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing