![Chlorotris(triphenylphosphine)cobalt(I) 97%](/deepweb/assets/sigmaaldrich/product/structures/228/160/b6712614-2e8d-4b79-af3b-7283d1c4fdd7/640/b6712614-2e8d-4b79-af3b-7283d1c4fdd7.png)
Chlorotris(triphenylphosphine)cobalt(I)
![Chlorotris(triphenylphosphine)cobalt(I) 97%](/deepweb/assets/sigmaaldrich/product/structures/228/160/b6712614-2e8d-4b79-af3b-7283d1c4fdd7/640/b6712614-2e8d-4b79-af3b-7283d1c4fdd7.png)
Linear Formula:
[(C6H5)3P]3CoCl
CAS Number:
Molecular Weight:
881.24
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing