Skip to Content
Merck
Chloroplatinic acid hydrate Technipur®, ≥99.9% trace metals basis

Chloroplatinic acid hydrate

Chloroplatinic acid hydrate Technipur®, ≥99.9% trace metals basis
Synonym(s):
Hexachloroplatinic(IV) acid hydrate, Hydrogen hexachloroplatinate(IV) hydrate, Platinic chloride hydrate
Linear Formula:
H2PtCl6 · xH2O
CAS Number:
Molecular Weight:
409.81 (anhydrous basis)

Products

Brand
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Core
Special Grade