Skip to Content
Merck
Chlorodibromoacetic acid analytical standard

Chlorodibromoacetic acid

Chlorodibromoacetic acid analytical standard
Synonym(s):
Dibromochloroacetic acid, Chlorodibromoacetic acid
Empirical Formula (Hill Notation):
C2HBr2ClO2
CAS Number:
Molecular Weight:
252.29

Products

Brand
Formula Weight
Melting Point (°C)
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing