Chlorobis(trimethylsilyl)methane
Synonym(s):
Bis(trimethylsilyl)chloromethane
Linear Formula:
[(CH3)3Si]2CHCl
CAS Number:
Molecular Weight:
194.85
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing