![Charcoal activated for the determination of AOX](/deepweb/assets/sigmaaldrich/product/images/288/300/26b922ce-5810-4461-b0b7-6309067d6d28/640/26b922ce-5810-4461-b0b7-6309067d6d28.jpg)
Charcoal activated
![Charcoal activated for the determination of AOX](/deepweb/assets/sigmaaldrich/product/images/288/300/26b922ce-5810-4461-b0b7-6309067d6d28/640/26b922ce-5810-4461-b0b7-6309067d6d28.jpg)
Synonym(s):
Charcoal activated
Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
Products
Brand
Formula Weight
Physical Form
Quality Segment
Application
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing