![Celiprolol for system suitability CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/391/165/77d0cbf4-f833-4778-9c29-96d9bed2895e/640/77d0cbf4-f833-4778-9c29-96d9bed2895e.png)
Celiprolol for system suitability
![Celiprolol for system suitability CRS, European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/391/165/77d0cbf4-f833-4778-9c29-96d9bed2895e/640/77d0cbf4-f833-4778-9c29-96d9bed2895e.png)
Synonym(s):
Celiprolol hydrochloride
Empirical Formula (Hill Notation):
C20H33N3O4 · HCl
CAS Number:
Molecular Weight:
415.95
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing