Skip to Content
Merck
(−)-Catechin phyproof® Reference Substance

(−)-Catechin

(−)-Catechin phyproof® Reference Substance
Synonym(s):
(−)-trans-3,3′,4′,5,7-Pentahydroxyflavane, (2S,3R)-2-(3,4-Dihydroxyphenyl)-3,4-dihydro-1(2H)-benzopyran-3,5,7-triol
Empirical Formula (Hill Notation):
C15H14O6
CAS Number:
Molecular Weight:
290.27
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Manufacturer
Physical Form
Product Line
Purity
Format
Application
Reference Material Type
Special Grade