![Cariprazine-D8 solution 100 μg/mL (Acetonitrile: Water with 1% 1M HCl (50:50) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/199/774/c17a1d34-43e3-41e5-b8dc-21bbe1087de3/640/c17a1d34-43e3-41e5-b8dc-21bbe1087de3.png)
Cariprazine-D8 solution
![Cariprazine-D8 solution 100 μg/mL (Acetonitrile: Water with 1% 1M HCl (50:50) (v/v)), certified reference material, ampule of 1 mL, Cerilliant®](/deepweb/assets/sigmaaldrich/product/structures/199/774/c17a1d34-43e3-41e5-b8dc-21bbe1087de3/640/c17a1d34-43e3-41e5-b8dc-21bbe1087de3.png)
Empirical Formula (Hill Notation):
C21H24D8Cl2N4O
CAS Number:
Molecular Weight:
435.46
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing