Carbasalate impurity B
Synonym(s):
2-[[2-(Acetyloxy)benzoyl]oxy]benzoic acid, 2-Carboxyphenyl o-acetylsalicylate
Empirical Formula (Hill Notation):
C16H12O6
CAS Number:
Molecular Weight:
300.26
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing