Cannabinoid Mixture (Acids) - 6 Component solution
Products
Brand
Manufacturer
Physical Form
Format
Chemical Composition
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing