Skip to Content
Merck
Calcium phosphate tribasic BioReagent, suitable for plant cell culture, powder

Calcium phosphate tribasic

Calcium phosphate tribasic BioReagent, suitable for plant cell culture, powder
Synonym(s):
Calcium hydroxyphosphate, HAp, Hydroxylapatite, Tribasic calcium phosphate
Linear Formula:
[Ca5(OH)(PO4)3]x
CAS Number:
Molecular Weight:
502.31
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing