Skip to Content
Merck
Buserelin acetate salt ≥90% (HPLC)

Buserelin acetate salt

Buserelin acetate salt ≥90% (HPLC)
Synonym(s):
des-Gly10-[D-Ser[t-Bu]6]-LH-RH ethylamide acetate salt, pGlu-His-Trp-Ser-Tyr-D-Ser(t-Bu)-Leu-Arg-Pro-NHEt acetate salt
Empirical Formula (Hill Notation):
C60H86N16O13 · C2H4O2
CAS Number:
Molecular Weight:
1299.48

Products

Brand
Formula Weight
Gene Alias
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing