
Bupropion Hydrochloride Related Compound B

Synonym(s):
2-(tert-Butylamino)-3′-bromopropiophenone hydrochloride
Empirical Formula (Hill Notation):
C13H18BrNO · HCl
CAS Number:
Molecular Weight:
320.65
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2