Skip to Content
AT
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Buffer solutions
Products
Brand
Supelco (3)
Product Line
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
certified reference material (3)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.99006
Buffer solutions,
traceable to NIST, traceable to PTB, pH 4.01 (25 °C, phthalate), pH 7.00 (25 °C, phosphate), pH 10.00(borate), Certipur
®
Expand
View Pricing
1.99005
Buffer solutions,
traceable to NIST, traceable to PTB, pH 4.01 (25 °C, phthalate), pH 7.00 (25 °C, phosphate), pH 9.00(borate), Certipur
®
Expand
View Pricing
1.99041
Buffer solutions,
(boric acid/potassium chloride/sodium hydroxide), traceable to SRM from NIST and PTB pH 11.00 (25°C) Certipur
®
Expand
View Pricing