Skip to Content
Merck
Bryostatin 1 ≥99%, solid

Bryostatin 1

Bryostatin 1 ≥99%, solid
Synonym(s):
Bryostatin 1
Empirical Formula (Hill Notation):
C47H68O17
CAS Number:
Molecular Weight:
905.03

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing