Skip to Content
Merck
(−)-Bornyl acetate phyproof® Reference Substance

(−)-Bornyl acetate

(−)-Bornyl acetate phyproof® Reference Substance
Synonym(s):
endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]hept-2-yl acetate
Empirical Formula (Hill Notation):
C12H20O2
CAS Number:
Molecular Weight:
196.29

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing