Skip to Content
Merck
Boc-Asp(OMe)-OH ≥98.0% (TLC)

Boc-Asp(OMe)-OH

Boc-Asp(OMe)-OH ≥98.0% (TLC)
Synonym(s):
Boc-L-aspartic acid 4-methyl ester
Linear Formula:
CH3O2CCH2CH[NHCO2C(CH3)3]CO2H
CAS Number:
Molecular Weight:
247.25

Products

Brand
Formula Weight
Physical Form
Purity
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing