Skip to Content
Merck
Bis(2,4-di-tert-butylphenyl) phosphate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Bis(2,4-di-tert-butylphenyl) phosphate

Bis(2,4-di-tert-butylphenyl) phosphate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Empirical Formula (Hill Notation):
C28H43O4P
CAS Number:
Molecular Weight:
474.61

Products

Brand
Formula Weight
Manufacturer
Physical Form
Quality Segment
Special Grade