
Bis(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate

Synonym(s):
Rhodium(I) tetrafluoroborate 1,5-Cyclooctadiene complex
Empirical Formula (Hill Notation):
C16H24BF4Rh
CAS Number:
Molecular Weight:
406.07
Products
Brand
Formula Weight
Melting Point (°C)
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reagent Type
Core
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing