Bilirubin Oxidase from Myrothecium verrucaria
Synonym(s):
Bilirubin:oxygen oxidoreductase
CAS Number:
Products
Brand
Sustainability
Biological Source
Feature
Physical Form
Chemical Composition
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing