Betaxolol for peak identification
Synonym(s):
Betaxolol hydrochloride, SL 75212
Empirical Formula (Hill Notation):
C18H29NO3 · HCl
CAS Number:
Molecular Weight:
343.89
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing