Skip to Content
Merck
BES ≥99.0% (titration)

BES

BES ≥99.0% (titration)
Synonym(s):
N,N-Bis(2-hydroxyethyl)-2-aminoethanesulfonic acid, N,N-Bis(2-hydroxyethyl)taurine
Empirical Formula (Hill Notation):
C6H15NO5S
CAS Number:
Molecular Weight:
213.25
EC Number:

Products

Product Category
Brand
Color
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Research Area
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing