Skip to Content
Merck
Benzyl alcohol, polymer-bound 70-90 mesh, extent of labeling: 2.0-3.0 mmol loading, 1 % cross-linked

Benzyl alcohol, polymer-bound

Benzyl alcohol, polymer-bound 70-90 mesh, extent of labeling: 2.0-3.0 mmol loading, 1 % cross-linked
Synonym(s):
(Hydroxymethyl)polystyrene, Benzyl alcohol polymer-bound, Hydroxymethyl polystyrene, Polystyrene cross-linked with divinylbenzene, hydroxymethylated

Products

Brand
Quality Segment
Functional Group
Cleavage Condition
Linker
Chain Type
Reaction Type