Benzene
Synonym(s):
Benzene
Empirical Formula (Hill Notation):
C6H6
CAS Number:
Molecular Weight:
78.11
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-7 of 7
Compare
Product Number
Description
Pricing