Skip to Content
Merck
Bensulfuron-methyl PESTANAL®, analytical standard

Bensulfuron-methyl

Bensulfuron-methyl PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C16H18N4O7S
CAS Number:
Molecular Weight:
410.40
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique