Skip to Content
Merck
Azoxystrobin certified reference material, TraceCERT®

Azoxystrobin

Azoxystrobin certified reference material, TraceCERT®
Empirical Formula (Hill Notation):
C22H17N3O5
CAS Number:
Molecular Weight:
403.39

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing