Azithromycin
Empirical Formula (Hill Notation):
C38H72N2O12
CAS Number:
Molecular Weight:
748.98
Products
Product Category
Brand
Formula Weight
Physical Form
Activity Spectrum
Research Area
Application
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing