Skip to Content
Merck
Azelastine hydrochloride European Pharmacopoeia (EP) Reference Standard

Azelastine Hydrochloride

Azelastine hydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Azelastine hydrochloride, 4-[(4-chlorophenyl)methyl]-2-(1-methylazepan-4-yl)phthalazin-1-one hydrochloride, Astelin, Optivar
Empirical Formula (Hill Notation):
C22H24ClN3O · HCl
CAS Number:
Molecular Weight:
418.36

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique