Skip to Content
Merck
Atrazine-desethyl PESTANAL®, analytical standard

Atrazine-desethyl

Atrazine-desethyl PESTANAL®, analytical standard
Synonym(s):
2-Amino-4-chloro-6-isopropylamino-1,3,5-triazine, Desethylatrazine
Empirical Formula (Hill Notation):
C6H10ClN5
CAS Number:
Molecular Weight:
187.63

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique