Skip to Content
Merck
Asiatic acid analytical standard

Asiatic acid

Asiatic acid analytical standard
Synonym(s):
2α,23-Dihydroxyursolic acid, Dammarolic acid, NSC 166063
Empirical Formula (Hill Notation):
C30H48O5
CAS Number:
Molecular Weight:
488.70
EC Number:

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4