Skip to Content
Merck
Apigenin 7-glucoside primary reference standard

Apigenin 7-glucoside

Apigenin 7-glucoside primary reference standard
Synonym(s):
4′,5,7-Trihydroxyflavone 7-glucoside, Apigenin-7-O-glucoside, Apigetrin, Cosmosiin, Apigenin 7-glucoside
Empirical Formula (Hill Notation):
C21H20O10
CAS Number:
Molecular Weight:
432.38
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique