![Anti-XRN1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/256/469/45ebb974-6c01-4cc1-9c77-215480887081/640/45ebb974-6c01-4cc1-9c77-215480887081.jpg)
Anti-XRN1 antibody produced in rabbit
![Anti-XRN1 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution](/deepweb/assets/sigmaaldrich/product/images/256/469/45ebb974-6c01-4cc1-9c77-215480887081/640/45ebb974-6c01-4cc1-9c77-215480887081.jpg)
Synonym(s):
Anti-40422, Anti-5′-3′ exoribonuclease 1, Anti-DKFZp434P0721, Anti-DKFZp686B22225, Anti-DKFZp686F19113, Anti-FLJ41903
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing