
Anti-SFPQ antibody, Mouse monoclonal

Synonym(s):
Monoclonal Anti-SFPQ antibody produced in mouse
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing