Skip to Content
Merck
Anti-RENBP antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Anti-RENBP antibody produced in rabbit

Anti-RENBP antibody produced in rabbit Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Synonym(s):
Anti-AGE antibody produced in rabbit, Anti-GlcNAc 2-epimerase antibody produced in rabbit, Anti-N-Acetyl-D-glucosamine 2-epimerase antibody produced in rabbit, Anti-N-Acylglucosamine 2-epimerase antibody produced in rabbit, Anti-Renin-binding protein antibody produced in rabbit, Anti-RnBP antibody produced in rabbit

Products

Product Category
Brand
Biological Source
Clonality
Feature
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique